missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6x-His Tag Monoclonal Antibody (HIS.H8), DyLight™ 550, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA121315D550
Additional Details : Weight : 0.01000kg
Description
MA1-21315-D550 has been successfully used for immunofluorescence. DyLight 550 has an excitation/emission of 562/576 nm.
The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombinant proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombinant protein using genetic engineering. By choosing a particular epitope and recombinant protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombinant proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent. His tag-specific antibodies are used to facilitate detection or coimmunopreciptation of tagged proteins.Specifications
6x-His Tag | |
Monoclonal | |
1 mg/mL | |
PBS with proprietary stabilizer and 0.02% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Tag | |
Antibody | |
IgG2b |
Immunocytochemistry, Western Blot | |
HIS.H8 | |
DyLight 550 | |
6His tag; 6X His; His Tag; Histidine Tag | |
6x His synthetic peptide. | |
50 μL | |
Primary | |
4° C, do not freeze | |
Liquid | |
Tag |