Learn More
Cytiva Cassette Rack
Brand:  Cytiva 80-6467-98
Additional Details : Weight : 2.05900kg
Product Code. 10462645
  • 5645.00 SEK / Pack of 2
Estimated Shipment: 28-05-2024
to see stock.
Specifications
Cassette Rack
Holds up to 14 gel cassettes
2 Pack