Learn More
Dissecting Set
Use this dissecting kit for a variety of applications. Dissecting Sets are suitable for theatre, ward, veterinary, dental and chiropody use.
Brand:  Samco Y261/74
Additional Details : Weight : 0.18000kg
Product Code. 13142141
  • 468.00 SEK / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
Stainless steel instruments in a wallet.
Kit includes:
  • 1 Scalpel, Polished Blade, SS Light Pattern, 35mm
  • 1 Scalpel, Polished Blade, SS Light Pattern, 45mm
  • 2 Dissecting Needles, all stainless steel
  • 1 PVC Dissecting Roll
  • 1 Spring Forceps Blunt, SS, 125mm
  • 1 Spring Forceps Sharp with Guide Pin, SS, 125mm
  • 1 Dissecting Scissors, Open Shanks, SS, 112mm
  • 1 Magnifier
Specifications
Specifications
Dissecting Set
Stainless Steel
1 Scalpel (35mm), 1 Scalpel (45mm), 2 Dissecting Needles, 1 PVC Dissecting Roll, 1 Spring Forceps Blunt (125mm), 1 Spring Forceps Sharp w/ Guide Pin (125mm), 1 Dissecting Scissors (Open Shanks, 112mm), 1 Magnifier