missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25420-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
May function as a transcriptional repressor. May also function as a ubiquitin ligase mediating ubiquitination of chromatin-associated TP53. Functions in cell survival and proliferation through control of the cell cycle. Functions in the p53 and pRB tumor suppressor pathways and regulates the cyclin CCNA2 transcription.; FUNCTION: Identified as a cellular target of the adenoviral oncoprotein E1A, it is required for both transcriptional activation and repression of viral genes.Specifications
DKC1 | |
Polyclonal | |
Unconjugated | |
DKC1 | |
CBF5, CBF5 homolog, DKC, DKC1, Dyskerin, NAP57, NOLA4, Nucleolar protein NAP57, snoRNP protein DKC1, XAP101 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1736 | |
-20°C | |
Liquid |
Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60832 | |
DKC1 | |
DKC1 Fusion Protein Ag7992 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |