Learn More
EVOS™ EVOS™ Condenser Slider, Meniscus B (semi-opaque liquids)
Even out field of view
Brand:  EVOS™ AMEPMN4
Additional Details : Weight : 2.00000kg
Product Code. 15369651
  • 455.00 SEK / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Used to improve illumination under certain conditions 
  • Due to variances in sample size, color and thickness, actual slider use may differ from suggestions
  • Meniscus A slider is intended for brightfield applications at 2x magnification with low-volume samples in multi-well plates and with clear liquids
  • Meniscus B slider is intended for brightfield applications at 2x magnification with high-volume samples in multi-well plates and with semi-opaque liquids
Specifications
Specifications
Meniscus B Condenser Slider
EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
1 Ea.
EVOS
Diffuser Slider, Pinhole Slider, Meniscus Sliders
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.