Learn More
Fisherbrand™ Heavy-Duty/Utility Funnels
Ideal for continuous lab work and heavy-duty jobs
168.48 SEK - 3230.00 SEK
Product Code | Brand | Capacity (Metric) | Diameter (Metric)Top | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric)Top | Height (Metric) | Price | Quantity & Availability | |||
12622445
|
Fisherbrand™
|
500 mL | 20.3 cm | 20.5 cm |
295.00 SEK
Pack of 3 |
In Stock
Log in to see stock. |
|||
11997934
|
Fisherbrand™
|
40 mL | 6.5cm | 9 cm |
1610.00 SEK
Pack of 72 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
11907944
|
Fisherbrand™
|
100 mL | 8.7cm | 11 cm |
1980.00 SEK
Pack of 72 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
11917914
|
Fisherbrand™
|
410 mL | 15.5cm | 17.7 cm |
2305.00 SEK
Pack of 24 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
11927944
|
Fisherbrand™
|
210 mL | 12.5cm | 14.1 cm |
3230.00 SEK
Pack of 72 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
11917944
|
Fisherbrand™
|
140 mL | 10.3cm | 11.5 cm |
1835.00 SEK
Pack of 72 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
12602445
|
Fisherbrand™
|
410 mL | 15.5cm | 17.7 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12662435
|
Fisherbrand™
|
40 mL | 6.5cm | 9 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12692435
|
Fisherbrand™
|
210 mL | 12.5cm | 14.1 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12682435
|
Fisherbrand™
|
100 mL | 8.7cm | 11 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12672435
|
Fisherbrand™
|
140 mL | 10.3cm | 11.5 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11997884
|
Fisherbrand™
|
500 mL | 20.3 cm | 20.5 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Ideal for continuous lab work and heavy-duty jobs
- Durable polypropylene construction with external ridges and ribbing to prevent vapor lock
- Autoclavable to 120°C (248°F)
Specifications
Polypropylene | |
Yes |