Learn More
Cole-Parmer™ Inspection Mirror
230.00 SEK - 474.00 SEK
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11729849
|
Cole-Parmer™
09914-00 |
31.75 mm |
230.00 SEK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
15119075
|
Cole-Parmer™
09914-05 |
47.625 mm |
474.00 SEK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
Specifications
1/Pk. | |
Inspection Mirror | |
Round |