Learn More
Proteintech Mouse Osteoprotegerin/TNFRSF11B ELISA Kit
Osteoprotegerin (OPG), also known as TNFRSF11B or OCIF, is a member of the tumor necrosis factor receptor super family (TNFRSF).
Brand:  Proteintech KE10058-96T
Product Code. 17244886
  • 3183.28 SEK / 96 reactions
Estimated Shipment: 13-11-2024
to see stock.
Explore more special offers
Description
It is a secreted glycoprotein that inhibits osteoclastogenesis and is involved in the regulation of bone density. Osteoprotegerin acts as decoy receptor for receptor activator of nuclear factor-kappa B ligand (RANKL) and neutralizes its function in osteoclastogenesis. Osteoprotegerin may function as a soluble decoy receptor for tumor necrosis factor (TNF)-related apoptosis inducing ligand (TRAIL) and inhibits TRAIL-induced apoptosis. TRAIL blocks osteoprotegerin mediated inhibition of osteoclastogenesis.
Specifications
Specifications
18383
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 96RXN Mouse Osteoprotegerin/TNFRSF11B ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine