missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOP14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26854-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Involved in nucleolar processing of pre-18S ribosomal RNA. Has a role in the nuclear export of 40S pre-ribosomal subunit to the cytoplasm.Specifications
NOP14 | |
Polyclonal | |
Unconjugated | |
NOP14 | |
C4orf9, NOL14, NOP14, Nucleolar complex protein 14, Nucleolar protein 14, RES4 25, RES425 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
289724, 75416, 8602 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P78316, Q8R3N1 | |
Nop14 | |
NOP14 Fusion Protein Ag25476 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |