Learn More
Fisherbrand™ Universal Replacement Shaft Bearing Seals
Fisherbrand™ Universal Replacement Shaft Bearing Seals are available in a range of sizes to fit different shaft diameters of most overhead stirrers.
Brand: Fisherbrand™
Additional Details : Weight : 0.01080kg
Description
- Universal replacement bearing seals
- Range of sizes available to fit different shaft diameters
- For overhead stirrers
Specifications
6 mm to 10 mm | |
Overhead Stirrers | |
Universal Replacement Bearing Seal |