Välkommen till Fisher Scientific
Buy 4 packs of Thermo Scientific Precise Protein Gelsget the Fisherbrand gel system for free
Thermo Scientific Precise Protein Gels are cast in a durable plastic cassette using a neutral pH buffer that prevents polyacrylamide breakdown and results in a long shelf life. Gels are individually packed in an easy-to-open plastic pouch and are ready to run with no comb or tape to remove. Fisherbrand Electrophoresis dual gel system (Verti-Gel mini) for precast gels incorporates a sealing system which is compatible with all major types of precast gels. The clamping system uses sliding locks which allows very rapid set up of the gels in the running unit. Ultra soft silicone seals and pressure bars which surround the cassette guarantee leak proof gel running. Casting accessories are not supplied but are available as optional extras.
Protein Ladders
Stained and Unstained Protein Ladders
Thermo Scientific Pierce Protein Ladders
Thermo Scientific Pierce protein ladder product line offers advanced quality and convenience for all your gel electrophoresis and Western blotting needs.
Highlights:
  • Cost effective – high quality without the high price performance – sharp protein-band resolution and consistent migration patterns provide easymolecule determination
  • Convenient – protein ladders are ready to load.
  • Reliable – exceptional lot-to-lot consistency and reproducibility
Select from the following ladders:
  • Thermo Scientific PageRuler Prestained Protein Ladder, 25µL
  • PageRuler® Plus Prestained Protein Ladder, 25µL
  • PageRuler Broad Range Unstained Protein Ladder, 25µL
  • SuperSignal® Molecular Weight Protein Ladder, 25µL
Item Number Product Description Bundle Price  
12334783 8-16% Precise Protein Gels, 10-Well 4x10 gels 4 880,00
12394783 4-20% Precise Protein Gels, 10-Well 4x10 gels 4 880,00
12314793 8-16% Precise Protein Gels, 12-Well 4x10 gels 4 880,00
12324793 4-20% Precise Protein Gels, 12-Well 4x10 gels 4 880,00
12344793 8-16% Precise Protein Gels, 15-Well 4x10 gels 4 880,00
12354793 4-20% Precise Protein Gels, 15-Well 4x10 gels 4 880,00
12364793 8-16%, Precise Protein Gels, 10-well, 10cm × 10cm 4x10 gels 4 880,00
12374793 4-20%, Precise Protein Gels, 10-well, 10cm × 10cm 4x10 gels 4 880,00
12384793 8-16%, Precise Protein Gels, 12-well, 10cm × 10cm 4x10 gels 4 880,00
12394793 4-20%, Precise Protein Gels, 12-well, 10cm × 10cm 4x10 gels 4 880,00
11533093 Vertical Gel tank Verti-gel Mini Precast Dual PAGE System with sliding clamps 0,00
11543110 PageRuler Prestained Protein Ladder 0,00
11573110 PageRuler Plus Prestained Protein Ladder 0,00
11553120 PageRuler Broad Range Unstained Protein Ladder 0,00
11547151 SuperSignal Enhanced Molecular Weight Protein Standards 0,00
Offer valid until 2012-12-31