CAS RN 1021462-91-8
CAS RN 1021462-91-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
(2R)-N-(2-Benzoyl-4-chlorophenyl)-1-(phenylmethyl)-2-pyrrolidinecarboxamide Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2R)-N-(2-Benzoyl-4-chlorophenyl)-1-(phenylmethyl)-2-pyrrolidinecarboxamide Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.