CAS RN 1033803-59-6
CAS RN 1033803-59-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
2-[(1RS)-3-(methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.