CAS RN 1044220-56-5
CAS RN 1044220-56-5
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
6-[2-Biotinylamidoethyl]-dithiopropionamido]-4,8-diaza-5,7-diketoundecanoic Acid Bis-N-sulfosuccinimidyl Ester Disodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-[2-Biotinylamidoethyl]-dithiopropionamido]-4,8-diaza-5,7-diketoundecanoic Acid Bis-N-sulfosuccinimidyl Ester Disodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6-[2-Biotinylamidoethyl]-dithiopropionamido]-4,8-diaza-5,7-diketoundecanoic Acid Bis-N-sulfosuccinimidyl Ester Disodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.