CAS RN 1047620-45-0
CAS RN 1047620-45-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
2-[(3a,4,5,6,7,7a-Hexahydro-1H-benzimidazol-2-yl)thio]-1-(4-morpholinyl)-ethanone Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-[(3a,4,5,6,7,7a-Hexahydro-1H-benzimidazol-2-yl)thio]-1-(4-morpholinyl)-ethanone Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.