CAS RN 104862-31-9
CAS RN 104862-31-9
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
[2R-(2a,3b,6a)]-3-Azido-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]octane-2-carboxylic Acid 5,5-Dioxide Diphenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
[2R-(2Alpha,3Beta,6Alpha)]-3-Azido-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]octane-2-carboxylic Acid 5,5-Dioxide Diphenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
[2R-(2Alpha,3Beta,6Alpha)]-3-Azido-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]octane-2-carboxylic Acid 5,5-Dioxide Diphenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.