CAS RN 1050506-87-0
CAS RN 1050506-87-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
GPR120 modulator 2, MedChemExpress
MedChemExpress GPR120 modulator 2 is a G protein coupled receptor 120 (GPR120) modulator extracted from patent US8394841B2, compound example F13. GPR120 modulator 2 can be used for the research of diseases associated with abnormal or deregulated GPR120, such as diabetes.