CAS RN 105102-18-9
CAS RN 105102-18-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Tibenelast sodium, MedChemExpress
MedChemExpress Tibenelast sodium is a phosphodiesterase inhibitor.
