CAS RN 106257-01-6
CAS RN 106257-01-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(16a,17b)- Estra-1,3,5(10)-triene-3,4,16,17-tetrol-16,17-diacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(16Alpha,17Beta)- Estra-1,3,5(10)-triene-3,4,16,17-tetrol-16,17-diacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(16Alpha,17Beta)- Estra-1,3,5(10)-triene-3,4,16,17-tetrol-16,17-diacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.