CAS RN 106862-82-2
CAS RN 106862-82-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
3-[(Tetrahydro-2H-pyran-2-yl)oxy]-19-Norpregna-1,3,5(10)-trien-20-yn-17-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-[(Tetrahydro-2H-pyran-2-yl)oxy]-19-Norpregna-1,3,5(10)-trien-20-yn-17-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-[(Tetrahydro-2H-pyran-2-yl)oxy]-19-Norpregna-1,3,5(10)-trien-20-yn-17-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.