accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1076-74-0

CH3OHNH3C

CAS RN 1076-74-0

IUPAC Name: 5-methoxy-2-methyl-1H-indole
Synonyms: 5-methoxy-2-methyl-1H-indole
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: VSWGLJOQFUMFOQ-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC(C)=CC2=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products