man

Test

CAS RN 1078-31-5

CH3SiH3CCH3OO

CAS RN 1078-31-5

IUPAC Name: 2-[(trimethylsilyl)oxy]benzaldehyde
Synonyms: 2-[(trimethylsilyl)oxy]benzaldehyde
Molecular Weight (g/mol): 194.31
Molecular Formula: C10H14O2Si
InChi Key: GMQJTDJLZSIFQG-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC1=CC=CC=C1C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results