CAS RN 1079-02-3
IUPAC Namn:
(²H₅)benzoic acid
Synonymer:
(²H₅)benzoic acid
Molekylvikt (g/mol):
127.15
Molekylär sammansättning:
C7H6O2
InChi Key:
WPYMKLBDIGXBTP-RALIUCGRSA-N
SMILES:
[2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C(O)=O
1
–
3
av
3
Resultat
1
Benzoic-d5 Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Benzoic Acid-d5, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Benzoic Acid-d5, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.