accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 108-50-9

CH3NH3CN

CAS RN 108-50-9

IUPAC Name: 2,6-dimethylpyrazine
Synonyms: 2,6-dimethylpyrazine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: HJFZAYHYIWGLNL-UHFFFAOYSA-N
SMILES: CC1=CN=CC(C)=N1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products