CAS RN 108078-14-4
CAS RN 108078-14-4
IUPAC Name:
2-iodo-3-methylbenzoic acid
Synonyms:
2-iodo-3-methylbenzoic acid
Molecular Weight (g/mol):
262.05
Molecular Formula:
C8H7IO2
InChi Key:
PZUXUOZSSYKAMX-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(C(O)=O)=C1I
1
–
1
of
1
results
1
2-Iodo-3-methylbenzoic acid, 98%, Thermo Scientific Chemicals
CAS: 108078-14-4 Molecular Formula: C8H7IO2 Molecular Weight (g/mol): 262.046 MDL Number: MFCD00079764 InChI Key: PZUXUOZSSYKAMX-UHFFFAOYSA-N Synonym: 2-iodo-m-toluic acid,benzoic acid,2-iodo-3-methyl,2-iodo-3-methyl-benzoic acid,pubchem12741,acmc-1bsuz,3-methyl-2-iodobenzoic acid,2-iodo-3-methyl benzoic acid,buttpark 148\07-11,2-iodo-3-methylbenzoic acid PubChem CID: 2759365 IUPAC Name: 2-iodo-3-methylbenzoic acid SMILES: CC1=CC=CC(=C1I)C(=O)O