accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1085-98-9

CH3NH3CSOONSFClCl

CAS RN 1085-98-9

IUPAC Name: ({[dichloro(fluoro)methyl]sulfanyl}(phenyl)sulfamoyl)dimethylamine
Synonyms: dichlofluanid
Molecular Weight (g/mol): 333.22
Molecular Formula: C9H11Cl2FN2O2S2
InChi Key: WURGXGVFSMYFCG-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)N(SC(F)(Cl)Cl)C1=CC=CC=C1
Solvent or Matrix 
  • (2)
  • (1)
Concentration 
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products