accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 109-05-7

CH3HN

CAS RN 109-05-7

IUPAC Name: 2-methylpiperidine
Synonyms: 2-methylpiperidine
Molecular Weight (g/mol): 99.18
Molecular Formula: C6H13N
InChi Key: NNWUEBIEOFQMSS-UHFFFAOYNA-N
SMILES: CC1CCCCN1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products