accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1092349-87-5

NH2BrFF

CAS RN 1092349-87-5

IUPAC Name: 2-bromo-3,4-difluoroaniline
Synonyms: 2-bromo-3,4-difluoroaniline
Molecular Weight (g/mol): 208.01
Molecular Formula: C6H4BrF2N
InChi Key: ZGZURPPCPVPDMX-UHFFFAOYSA-N
SMILES: NC1=C(Br)C(F)=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products