man

Test

CAS RN 1092461-25-0

ClNOFFF

CAS RN 1092461-25-0

IUPAC Name: 5-chloro-2-(trifluoromethoxy)benzonitrile
Synonyms: 5-chloro-2-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 221.56
Molecular Formula: C8H3ClF3NO
InChi Key: MBALELAICTYSDI-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=C(C=C(Cl)C=C1)C#N

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results