CAS RN 1092461-31-8
CAS RN 1092461-31-8
IUPAC Name:
3-ethoxy-2,6-difluorophenol
Synonyms:
3-ethoxy-2,6-difluorophenol
Molecular Weight (g/mol):
174.15
Molecular Formula:
C8H8F2O2
InChi Key:
MGBIYPQQWRPKLL-UHFFFAOYSA-N
SMILES:
CCOC1=C(F)C(O)=C(F)C=C1
1
–
1
of
1
results
1
3-Ethoxy-2,6-difluorophenol, 97%, Thermo Scientific™
CAS: 1092461-31-8 Molecular Formula: C8H8F2O2 Molecular Weight (g/mol): 174.147 MDL Number: MFCD11519373 InChI Key: MGBIYPQQWRPKLL-UHFFFAOYSA-N Synonym: 3-ethoxy-2,6-difluoro-phenol PubChem CID: 55275768 IUPAC Name: 3-ethoxy-2,6-difluorophenol SMILES: CCOC1=C(C(=C(C=C1)F)O)F