accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1093860-48-0

CH3OONNCl

CAS RN 1093860-48-0

IUPAC Name: methyl 6-chloropyridazine-4-carboxylate
Synonyms: methyl 6-chloropyridazine-4-carboxylate
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: QYQHGLKSLGUINZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=NN=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products