accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1095-03-0

OBOO

CAS RN 1095-03-0

IUPAC Name: triphenyl borate
Synonyms: boric acid, triphenyl ester
Molecular Weight (g/mol): 290.13
Molecular Formula: C18H15BO3
InChi Key: MDCWDBMBZLORER-UHFFFAOYSA-N
SMILES: O(B(OC1=CC=CC=C1)OC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products