CAS RN 109518-47-0
CAS RN 109518-47-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
2-[(1RS)-8-Ethyl-1-methyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (1-Methyl Etodolac), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.