accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1095382-05-0

CH3NONNNBrNHNNNH3C

CAS RN 1095382-05-0

IUPAC Name: 1-{6-bromo-2-[4-(4-methylpiperazin-1-yl)phenyl]-1H-imidazo[4,5-b]pyridin-7-yl}-4-[(5-methyl-1,2-oxazol-3-yl)methyl]piperazine
Synonyms: 1-{6-bromo-2-[4-(4-methylpiperazin-1-yl)phenyl]-1H-imidazo[4,5-b]pyridin-7-yl}-4-[(5-methyl-1,2-oxazol-3-yl)methyl]piperazine
Molecular Weight (g/mol): 551.49
Molecular Formula: C26H31BrN8O
InChi Key: GFLQCBTXTRCREJ-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=CC=C(C=C1)C1=NC2=NC=C(Br)C(N3CCN(CC4=NOC(C)=C4)CC3)=C2N1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products