CAS RN 110347-85-8
CAS RN 110347-85-8
IUPAC Name:
(2S,4R)-4-(phosphonomethyl)piperidine-2-carboxylic acid
Synonyms:
selfotel
Molecular Weight (g/mol):
223.17
Molecular Formula:
C7H14NO5P
InChi Key:
LPMRCCNDNGONCD-RITPCOANSA-N
SMILES:
OC(=O)[C@@H]1C[C@H](CP(O)(O)=O)CCN1
1
–
1
of
1
results
1
CGS 19755, Tocris Bioscience™
CAS: 110347-85-8 Molecular Formula: C7H14NO5P Molecular Weight (g/mol): 223.165 InChI Key: LPMRCCNDNGONCD-RITPCOANSA-N Synonym: selfotel,selfotel usan:inn,2s,4r-4-phosphonomethyl piperidine-2-carboxylic acid,cis-4-phosphonomethyl pipecolic acid,chembl39664,2-piperidinecarboxylic acid, 4-phosphonomethyl-, cis,2-piperidinecarboxylic acid, 4-phosphonomethyl-, 2s,4r,cis-4-phosphonomethyl piperidine-2-carboxylic acid,cis-4-phosphonomethyl-piperidine-2-carboxylic acid,selfotel, + PubChem CID: 68736 ChEBI: CHEBI:34973 IUPAC Name: (2S,4R)-4-(phosphonomethyl)piperidine-2-carboxylic acid SMILES: C1CNC(CC1CP(=O)(O)O)C(=O)O