accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 110919-71-6

CH3NH2NF

CAS RN 110919-71-6

IUPAC Name: 5-fluoro-6-methylpyridin-2-amine
Synonyms: 5-fluoro-6-methylpyridin-2-amine
Molecular Weight (g/mol): 126.13
Molecular Formula: C6H7FN2
InChi Key: WMGRXNKQEVOVET-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC(N)=N1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products