accessibility menu, dialog, popup

UserName

CAS RN 111-48-8

HOSOH

CAS RN 111-48-8

IUPAC Name: 2-[(2-hydroxyethyl)sulfanyl]ethan-1-ol
Synonyms: thiodiglycol
Molecular Weight (g/mol): 122.18
Molecular Formula: C4H10O2S
InChi Key: YODZTKMDCQEPHD-UHFFFAOYSA-N
SMILES: OCCSCCO
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products