accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 11120-25-5

NH4NH4OWOOO

CAS RN 11120-25-5

IUPAC Name: diammonium dioxotungstenbis(olate)
Synonyms: ammonium paratungstate
Molecular Weight (g/mol): 283.91
Molecular Formula: H8N2O4W
InChi Key: IZYSFFFJIOTJEV-UHFFFAOYSA-P
SMILES: [NH4+].[NH4+].[O-][W]([O-])(=O)=O
Grade 
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)
Technique 
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products
1x1 image pixel for data collection