CAS RN 111385-66-1
CAS RN 111385-66-1
IUPAC Name:
2-(2,3,4,5,6-pentamethylbenzoyl)benzoic acid
Synonyms:
2-(2,3,4,5,6-pentamethylbenzoyl)benzoic acid
Molecular Weight (g/mol):
296.37
Molecular Formula:
C19H20O3
InChi Key:
DQCUMOMIHACOJZ-UHFFFAOYSA-N
SMILES:
CC1=C(C)C(C)=C(C(=O)C2=CC=CC=C2C(O)=O)C(C)=C1C
1
–
1
of
1
results
1
2-(2,3,4,5,6-Pentamethylbenzoyl)benzoic acid, 95%, Thermo Scientific™
CAS: 111385-66-1 Molecular Formula: C19H20O3 Molecular Weight (g/mol): 296.366 MDL Number: MFCD00051629 InChI Key: DQCUMOMIHACOJZ-UHFFFAOYSA-N Synonym: 2-2,3,4,5,6-pentamethylbenzoyl benzoic acid,benzoic acid,2-2,3,4,5,6-pentamethylbenzoyl,2-pentamethylbenzoylbenzoic acid,2-pentamethylbenzoyl benzoic acid,maybridge1_002905,acmc-1br1r,2-2,3,4,5,6-pentamethylphenyl carbonylbenzoic acid,2-2,3,4,5,6-pentamethylphenyl carbonyl benzoic acid,2-oxo-2,3,4,5,6-pentamethylphenyl methyl benzoic acid PubChem CID: 727751 IUPAC Name: 2-(2,3,4,5,6-pentamethylbenzoyl)benzoic acid SMILES: CC1=C(C(=C(C(=C1C)C)C(=O)C2=CC=CC=C2C(=O)O)C)C