accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 11140-77-5

NH3NH3NH3NH3NH3NH3NH3NH3NH3NH3OHWOOWOOWOOOWOOWOOOWOOWOOOWOOOWOOOWOOWOOOWOOOHOOHOHHOHOHOHOOHOH

CAS RN 11140-77-5

IUPAC Name: dodecaoxooctacyclo[21.1.1.1¹,³.1³,⁵.1⁷,⁹.1¹¹,¹³.1¹⁵,¹⁷.1¹⁹,²¹]dodecatungstoxane-5,7,9,11,13,15,17,19,21,23-decol decaamine
Synonyms: dodecaoxooctacyclo[21.1.1.1¹,³.1³,⁵.1⁷,⁹.1¹¹,¹³.1¹⁵,¹⁷.1¹⁹,²¹]dodecatungstoxane-5,7,9,11,13,15,17,19,21,23-decol decaamine
Molecular Weight (g/mol): 3042.43
Molecular Formula: H40N10O41W12
InChi Key: LMAJGSPZJYEENT-UHFFFAOYSA-D
SMILES: N.N.N.N.N.N.N.N.N.N.O[W]12(=O)O[W](O)(=O)(O1)O[W]1(O)(=O)O[W](O)(=O)(O1)O[W]1(O)(=O)O[W]3(=O)(O1)O[W]1(=O)(O[W](O)(=O)(O1)O[W]1(O)(=O)O[W](O)(=O)(O1)O[W]1(O)(=O)O[W](O)(=O)(O1)O2)O3
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products