CAS RN 1119-33-1
CAS RN 1119-33-1
IUPAC Name:
2-amino-5-ethoxy-5-oxopentanoic acid
Synonyms:
5-ethyl-L-glutamate
Molecular Weight (g/mol):
175.18
Molecular Formula:
C7H13NO4
InChi Key:
XMQUEQJCYRFIQS-UHFFFAOYNA-N
SMILES:
CCOC(=O)CCC(N)C(O)=O
1
–
1
of
1
results
1
L-Glutamic acid 5-ethyl ester, 95%, Thermo Scientific™
CAS: 1119-33-1 Molecular Formula: C7H13NO4 Molecular Weight (g/mol): 175.18 MDL Number: MFCD00037217 InChI Key: XMQUEQJCYRFIQS-UHFFFAOYNA-N Synonym: h-glu oet-oh,l-glutamic acid 5-ethyl ester,l-glutamic acid gamma-ethyl ester,ethyl glutamate,unii-3ec85nwr4k,5-ethyl-l-glutamate,s-2-amino-5-ethoxy-5-oxopentanoic acid,2s-2-amino-5-ethoxy-5-oxopentanoic acid,3ec85nwr4k,l-glutamic acid-5-ethyl ester PubChem CID: 3082305 SMILES: CCOC(=O)CCC(N)C(O)=O