accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112-59-4

H3COOOH

CAS RN 112-59-4

IUPAC Name: 2-[2-(hexyloxy)ethoxy]ethan-1-ol
Synonyms: 2-[2-(hexyloxy)ethoxy]ethanol
Molecular Weight (g/mol): 190.28
Molecular Formula: C10H22O3
InChi Key: GZMAAYIALGURDQ-UHFFFAOYSA-N
SMILES: CCCCCCOCCOCCO
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products