accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1122-41-4

SClCl

CAS RN 1122-41-4

IUPAC Name: (2,4-dichlorophenyl)sulfanide
Synonyms: (2,4-dichlorophenyl)sulfanide
Molecular Weight (g/mol): 178.05
Molecular Formula: C6H3Cl2S
InChi Key: FGBVJFREPSJSNG-UHFFFAOYSA-M
SMILES: [S-]C1=CC=C(Cl)C=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products