accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1123-86-0

CH3O

CAS RN 1123-86-0

IUPAC Name: 1-cyclohexylpropan-1-one
Synonyms: 1-cyclohexyl-propan-1-one
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: AMHOPTNGSNYSBL-UHFFFAOYSA-N
SMILES: CCC(=O)C1CCCCC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products