accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1124-64-7

H3CN

CAS RN 1124-64-7

IUPAC Name: 1-butylpyridin-1-ium
Synonyms: 1-butylpyridin-1-ium
Molecular Weight (g/mol): 136.22
Molecular Formula: C9H14N
InChi Key: REACWASHYHDPSQ-UHFFFAOYSA-N
SMILES: CCCC[N+]1=CC=CC=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products