accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1125-80-0

H3CN

CAS RN 1125-80-0

IUPAC Name: 3-methylisoquinoline
Synonyms: isoquinolone-3-methyl
Molecular Weight (g/mol): 143.19
Molecular Formula: C10H9N
InChi Key: FVVXWRGARUACNW-UHFFFAOYSA-N
SMILES: CC1=NC=C2C=CC=CC2=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products