CAS RN 1125410-09-4
CAS RN 1125410-09-4
IUPAC Name:
3-iodopyridine-2,5-diamine
Synonyms:
3-iodopyridine-2,5-diamine
Molecular Weight (g/mol):
235.03
Molecular Formula:
C5H6IN3
InChi Key:
SECWUDQFVZPDOS-UHFFFAOYSA-N
SMILES:
NC1=CC(I)=C(N)N=C1