accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112780-04-8

CH3OOH3COH

CAS RN 112780-04-8

IUPAC Name: methyl 4-hydroxyhex-2-ynoate
Synonyms: methyl 4-hydroxyhex-2-ynoate
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: GMESNUCOCRRYJV-UHFFFAOYNA-N
SMILES: CCC(O)C#CC(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products