accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1128-00-3

H3COONH2

CAS RN 1128-00-3

IUPAC Name: ethyl 2-aminocyclohex-1-ene-1-carboxylate
Synonyms: ethyl 2-aminocyclohex-1-ene-1-carboxylate
Molecular Weight (g/mol): 169.22
Molecular Formula: C9H15NO2
InChi Key: JBZVWABPSHNPIK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)CCCC1
Physical Form 
  • (1)
Quantity 
  • (1)
1x1 image pixel for data collection 1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products