accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1131-87-9

SOBr

CAS RN 1131-87-9

IUPAC Name: 1-(1-benzothiophen-5-yl)-2-bromoethan-1-one
Synonyms: 1-(1-benzothiophen-5-yl)-2-bromoethanone
Molecular Weight (g/mol): 255.13
Molecular Formula: C10H7BrOS
InChi Key: NTQPGUCRHJHYIA-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C2SC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products